Recombinant Pig CXCL8 (from E. coli)
Catalog # ABNOP9481.20UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Pig
- Gene ID:P26894
- Protein/peptide name:CXCL8
- Sequence:26a.a.-103a.a.