0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Proteins and Peptides
Recombinant Human CXCL8 (from E. coli)
Recombinant Human CXCL8 (from E. coli)
ABNOP9482.25UG
:
Abnova
:
Recombinant Human CXCL8 (from E. coli)
ABNOP9482.25UG
:
Abnova
:
Pk:
25 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Human
Gene ID:
P10145
Protein/peptide name:
CXCL8
Sequence:
23a.a.-99a.a.