Order Entry
Denmark
ContactUsLinkComponent
Recombinant Human CXCL8 (from P. pastoris)
Recombinant Human CXCL8 (from P. pastoris)
Catalog # ABNOP9483.25UG
Supplier:  Abnova
CAS Number:  
Recombinant Human CXCL8 (from P. pastoris)
Catalog # ABNOP9483.25UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    P. pastoris
  • Species:
    Human
  • Gene ID:
    P10145
  • Protein/peptide name:
    CXCL8

Specifications

About this item