Recombinant Human IFNGR2 (from CHO cells)
Catalog # ABNOP7377.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Gene ID:P38484
- Protein/peptide name:IFNGR2
- Sequence:28a.a.-247a.a