Recombinant Human IL11 (from CHO cells)
Catalog # ABNOP7097.5UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:5 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Gene ID:P202809-1
- Protein/peptide name:IL11
- Sequence:22a.a.-199a.a.