Recombinant Human PDFGB (from P. pastoris)
Catalog # ABNOP7140.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:P. pastoris
- Species:Human
- Gene ID:P01127
- Protein/peptide name:PDFGB
- Sequence:82a.a.-190a.a.