Order Entry
Denmark
ContactUsLinkComponent
Recombinant Mouse VEGFA (from P. pastoris)
Recombinant Mouse VEGFA (from P. pastoris)
Catalog # ABNOP7146.1MG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse VEGFA (from P. pastoris)
Catalog # ABNOP7146.1MG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    P. pastoris
  • Species:
    Mouse
  • Gene ID:
    Q00731-2
  • Protein/peptide name:
    VEGFA
  • Sequence:
    27a.a.-190a.a.

Specifications

About this item