Recombinant Human CSF2 (from P. pastoris)
Catalog # ABNOP7149.1MG
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:P. pastoris
- Species:Human
- Gene ID:P04141
- Protein/peptide name:CSF2
- Sequence:18a.a.-144a.a.