Order Entry
Denmark
ContactUsLinkComponent
Anti-Fructose 6 Phosphate Kinase Rabbit Recombinant Antibody [clone: R08-6B4]
Anti-Fructose 6 Phosphate Kinase Rabbit Recombinant Antibody [clone: R08-6B4]
Catalog # ABNORAB01718.100UL
Supplier:  Abnova
Anti-Fructose 6 Phosphate Kinase Rabbit Recombinant Antibody [clone: R08-6B4]
Catalog # ABNORAB01718.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    Fructose 6 Phosphate Kinase
  • Clonality:
    Recombinant
  • Clone:
    R08-6B4
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Fructose 6 Phosphate Kinase
  • Pk:
    100 µl

Specifications

About this item