Order Entry
Denmark
ContactUsLinkComponent
Anti-MGMT Rabbit Recombinant Antibody [clone: R01-8B1]
Anti-MGMT Rabbit Recombinant Antibody [clone: R01-8B1]
Catalog # ABNORAB01747.100UL
Supplier:  Abnova
Anti-MGMT Rabbit Recombinant Antibody [clone: R01-8B1]
Catalog # ABNORAB01747.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    MGMT
  • Clonality:
    Recombinant
  • Clone:
    R01-8B1
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of human MGMT
  • Pk:
    100 µl

Specifications

About this item