Anti-GABARAPL2 Rabbit Recombinant Antibody [clone: R03-7J2]
Catalog # ABNORAB02268.100UL
Supplier: Abnova
Specifications
- Antigen symbol:GABARAPL2
- Clonality:Recombinant
- Clone:R03-7J2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human GABARAPL2.
- Pk:100 µl