Anti-FCGR1A Rabbit Recombinant Antibody [clone: R08-6A8]
Catalog # ABNORAB02283.100UL
Supplier: Abnova
Specifications
- Antigen symbol:FCGR1A
- Clonality:Recombinant
- Clone:R08-6A8
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human FCGR1A.
- Pk:100 µl