Anti-RB1 (pSer807/S811) Rabbit Monoclonal Antibody [clone: D9] (APC)
Catalog # ABNOMAB23426.100
Supplier: Abnova
Specifications
- Antigen symbol:RB1
- Clonality:Monoclonal
- Clone:D9
- Conjugation:APC (Allophycocyanin)
- Host:Rabbit
- Reactivity:Human
- Modification:pSer807/S811
- Immunogen:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human RB1
- Pk:100 AU