Order Entry
Denmark
Orders LinkContactUsLinkComponent
Square Containers

Square Containers

100 ml, 87×68×26 mm
Catalog # CERB117170
Supplier:   NOLATO HERTILA
Square Containers
Catalog # CERB117170
Supplier:  NOLATO HERTILA
Square Containers

100 ml, 87×68×26 mm

Price
30.440,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents

Square container made of polypropylene.

Specifications
  • Capacity:
    100 ml
  • W×D×H:
    87×68×26 mm