Order Entry
Denmark
Orders LinkContactUsLinkComponent
Halogen lamp 24 V

Halogen lamp 24 V

B15s, 24 V, 21 V W
Catalog # 139-0146
Supplier:   OSRAM
Halogen lamp 24 V
Catalog # 139-0146
Supplier:  OSRAM
Halogen lamp 24 V

B15s, 24 V, 21 V W

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

Type of bulb commonly used in lorries/trucks.

Specifications
  • Base:
    B15s
  • Power:
    21 V W
  • Voltage:
    24 V
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.