Order Entry
Denmark
ContactUsLinkComponent
Experimentation kits, cleavage of Lambda DNA with Eco RI endonuclease: intro to restriction enzymes
Experimentation kits, cleavage of Lambda DNA with Eco RI endonuclease: intro to restriction enzymes
Catalog # EDVO212
Supplier:  EDVOTEK
undefined
Experimentation kits, cleavage of Lambda DNA with Eco RI endonuclease: intro to restriction enzymes
Catalog # EDVO212
Supplier:  EDVOTEK
Specifications
  • Description:
    Cleavage of Lambda DNA with Eco RI Endonuclease: Intro to Restriction Enzymes
  • Pk:
    1 KIT
Specifications
About this item
The DNA from bacteriophage Lambda is a well-characterised linear molecule containing six recognition sites for Eco RI (5 distinct sites, 2 are very close in size). In this experiment, Lambda DNA is digested by the Eco RI endonuclease. The digestion products are analysed by agarose gel electrophoresis.
  • For 5 lab groups
  • Complete in 90 minutes
Also required: Electrophoresis tank, power supply, 5 to 50 µl and 40 µl fixed volume micropipettes, water bath, and white light box.
Delivery: Kit includes instructions, Lambda DNA, Dryzymes™, reconstitution buffer, restriction enzyme reaction buffer, enzyme grade water, standard DNA fragments, various solutions and buffers, agarose powder, and stains.