Order Entry
Denmark
Orders LinkContactUsLinkComponent
Liquid Dip Samplers
Liquid Dip Samplers
Liquid Dip Samplers
Liquid Dip Samplers
Liquid Dip Samplers
The stainless steel liquid dip sampler is used to collect a wide variety of liquid samples from the surface.
  • Made from high quality and chemically resistant 316L stainless steel
  • Different sizes available
  • Convenient hands-free emptying feature
Once the sample has been collected, the liquid dip sampler can simply be pushed against the container allowing for the liquid to be easily emptied.
Product Family Options
  • Description
    Material
    Capacity
  • Liquid Dip sampler
    Stainless steel 316L
    50 ml
    1 * 1 Items
    1 * 1 Items - 2.630,00 kr
    2.630,00 kr
    1 * 1 Items
       2.630,00 kr
  • Liquid Dip sampler
    Stainless steel 316L
    100 ml
    1 * 1 Items
    1 * 1 Items - 2.980,00 kr
    2.980,00 kr
    1 * 1 Items
       2.980,00 kr