Analytical Membrane Bands
Analytical membrane bands are perfect for both microbial quality assurance (QA) and quality control (QC) applications where membrane dispensers are used.
- Membranes made of MCE material
- Bands are numbered to indicate how many membranes remain to be dispensed
- Membranes are gridded to improve accuracy and save time
Compatible with most common dispensers on the market, including MilliporeSigma™ and Sartorius™, giving added flexibility and security of supply. Boxes feature two marked orientations and tear-away perforations with a thumb-tab for quick and easy loading.
: Product has been validated to the highest standards, including ISO 7704 and ASTM F838.
: Each case of pre-sterilised analytical membrane bands contains four boxes of 150 membranes per pack (600/case total).
- Catalog No:
- 514-1431
- 514-1430
- 514-1435
- 514-1433
- 514-1432
- 514-1434
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together