Order Entry
Denmark
ContactUsLinkComponent
Screw vials, Thermo Scientific™ Chromacol™
Screw vials, Thermo Scientific™ Chromacol™
Catalog # HYPE1.1-STVGN
Screw vials, Thermo Scientific™ Chromacol™
Catalog # HYPE1.1-STVGN
Specifications
  • Cap Size:
    8-425
  • Ø×H:
    12×32 mm
  • Capacity:
    1,10 ml
  • Colour:
    Clear
  • Description:
    Screw vial
  • Pk:
    500
Specifications
About this item
Thermo Scientific™ Chromacol™ sample storage screw thread vials are manufactured from superior quality 33 expansion borosilicate glass, clear (Type 1, Class A).
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample.