Order Entry
Denmark
ContactUsLinkComponent
SPE Plate, 96-well, NBE OFQC
SPE Plate, 96-well, NBE OFQC
Catalog # TCAT637-0051NBE
Supplier:  Tecan
SPE Plate, 96-well, NBE OFQC
Catalog # TCAT637-0051NBE
Supplier:  Tecan

Specifications

  • No. of wells:
    96
  • Description:
    Plate NBE OFQC 1 ml 30 µm
  • Environmentally Preferable:
  • Phase:
    OFQC
  • Filling:
    5 mg
  • Pk:
    1

Specifications

About this item

A proprietary polymer resin blend of ion-exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.