Order Entry
Denmark
ContactUsLinkComponent
 
SPE Plate, 96-well, NBE HPSAX
  TCAT412-0051NBE
 :  Tecan
SPE Plate, 96-well, NBE HPSAX
  TCAT412-0051NBE
 :  Tecan

 

  • No. of wells:
    96
  • Description:
    Plate NBE HPSAX, 1 ml, 20 µm
  • Environmentally Preferable:
  • Filling:
    5 mg
  • Pk:
    1

 

 

Polymeric strong anionic exchange chemistry.

  • Ion-exchange
  • Liquid handler compatible

20 µm polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in fast and uniform sample prep. Ideal for acidic compounds.