Order Entry
Denmark
ContactUsLinkComponent
SPE Column, NBE WWP2
SPE Column, NBE WWP2
Catalog # TCAT417-0051R-NBE
Leverandør:  Tecan
SPE Column, NBE WWP2
Catalog # TCAT417-0051R-NBE
Leverandør:  Tecan

Specifikationer

  • Particle Size:
    30 µm
  • Pore size:
    300 Å
  • Capacity:
    1 ml
  • Description:
    Column NBE WWP2
  • Bed weight:
    5 mg
  • Phase:
    WWP2
  • Pk:
    96

Specifikationer

Om denne vare

Polymeric water-wettable resin ideal for reverse-phase retention of aqueous samples.

  • Reversed-phase
  • Liquid handler compatible

Special water wettable polymer with 300 Å pore size ideal for protein digest clean up and other protein related workflows.