Order Entry
Denmark
Orders LinkContactUsLinkComponent
Relay Module
Relay Module
Relay module potential-free forwarding of 4 different alarm states
Catalog # MSGA38766
Supplier:   asecos
Relay Module
Catalog # MSGA38766
Supplier:  asecos
Relay Module
Relay Module
Relay module potential-free forwarding of 4 different alarm states
Price
1.660,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
By choosing this option, you are opting for alarm differentiation instead of a collective alarm.
These signals are passed on to a central control centre via potential-free alarm contacts. With the clear differentiation of the alarm conditions, you can initiate optimally coordinated measures.
The relay module is suited for the PRO models of the IONLINE.
Specifications
  • Description:
    Relay module potential-free forwarding of 4 different alarm states
  • Seq:
    166
Anbefalinger