Order Entry
Denmark
Orders LinkContactUsLinkComponent
Potassium hydroxide

Potassium hydroxide

50 mL
Katalog # SUPLP4494-50ML
Leverandør:   Merck
Potassium hydroxide
Katalog # SUPLP4494-50ML
Leverandør:  Merck
CAS nummer:  1310-58-3
Potassium hydroxide

50 mL

CAS nummer:  1310-58-3
Price
752,00 kr
Log in for availability
1 * 50 mL
Product Details & Documents
Synonyms:  Potash caustic, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol
Specifications

Specifications

  • Pk:
    50 mL

Specifications