FectoVIR®-LV Transfection Reagents
Supplier: Polyplus-transfection
Certificates
About this item
Transfection reagents for lentiviral vector production from small to large scale.
- Productivity: Reach highest lentiviral vectors titers in suspension systems
- Cost-effectiveness: Reduce your cost per batch with high titers and low DNA consumption
- Scalability: Produce at large scale with low complexation volume and long complex stability
- Time saving: Use polyplus DOE service to optimize your process in a record time
FectoVIR®-LV is the next generation of transfection reagent, free of animal component, designed to improve Lentiviral vector productivity in HEK-293 cell systems.
FectoVIR®-LV is made for large scale manufacturing with a reduction of the complexation volume and increased complex stability.
The FectoVIR-LV benefits allow to increase the number of doses produced per batch, while decreasing LVV manufacturing costs.
Certifications: ISO 9001 facility certifications.
Specifications
- Bioproduction:Yes
- Cell specific:HEK-293 and derivatives
- DNA:Yes
- Virus production:Yes
- Storage temperature:5 ±3 °C
Specifications
Frequently Bought Together