VWR® Vial Grippers
Leverandør: VWR Collection
Certifikater
Om denne vare
These two styles of hand-operated grippers are designed to pick up cryogenic and crimped 10R-size vaccine/serum vials, respectively.
- Each style is available in a pack of five assorted colours (red, blue, green, purple, yellow) for easy identification and organisation
- A combo pack with one of each style is also available
- Reinforced polypropylene
- Autoclavable
- Reusable
- Plastic-free packaging
Both are durably made of reinforced polypropylene which is cold-tolerant and resistant to everyday wear. The designated squeeze target ensures maximum precision and grip with every use.
Frequently Bought Together