Rubis® Cutting Tweezers
Leverandør: OUTILS RUBIS
Certifikater
Om denne vare
Cutting tweezers with small head for cutting soft material or with large head for cutting strong material.
- Made from DURAX hardened stainless steel
- Not autoclavable
- Easy to clean
High chemical resistance, temperature resistant up to 400 °C.
Frequently Bought Together