SlimT™ Mop Covers
About this item
Low-profile and economical, rounded T-mop covers that feature snaps on one side and a padded slip on the other to easily clean windows, awkward corners, tight spaces, and under equipment. Available in a range of fabrics for specialized cleaning.
- Low profile, rounded covered sponges
- Economical design for wide cleanroom use
- Padded end protects equipment while cleaning
- Excellent on all room surfaces and equipment exteriors
- Easily fits in awkward corners, tight spaces and under equipment
- 304,8 and 457,2 mm options are ideal for use with the QPWT series window washing tool
- Holds up to strong disinfectants
- Autoclavable
Selection of fabrics include:
NovaPoly™ - 100% smooth knit polyester provides excellent disinfectant dispersal
PolySorb™ - 100% textured polyester for removing residues and dispersing disinfectant
MegaTex™ - 100% non-woven polyamide is absorbent with a mildly abrasive surface
Microfiber - 80% polyester, 20% polyamide is highly absorbent and effective wet or dry
The Micronova Slim T™ Mop Covers fit onto the 180° swivel T-Mop™ adapter. Available with an optional thicker foam layer to hold more disinfectant and facilitate wringing. The 305 and 457 mm options are designed for use with the micronova window washing tool for a streak-free finish.
: All components must be ordered separately. For additional Universal Handle Lengths, see Universal Mop Handles from Micronova.
9 Options Available Below
Product Details & Documents
Low-profile and economical, rounded T-mop covers that feature snaps on one side and a padded slip on the other to easily clean windows, awkward corners, tight spaces, and under equipment. Available in a range of fabrics for specialized cleaning.
- Low profile, rounded covered sponges
- Economical design for wide cleanroom use
- Padded end protects equipment while cleaning
- Excellent on all room surfaces and equipment exteriors
- Easily fits in awkward corners, tight spaces and under equipment
- 304,8 and 457,2 mm options are ideal for use with the QPWT series window washing tool
- Holds up to strong disinfectants
- Autoclavable
Selection of fabrics include:
NovaPoly™ - 100% smooth knit polyester provides excellent disinfectant dispersal
PolySorb™ - 100% textured polyester for removing residues and dispersing disinfectant
MegaTex™ - 100% non-woven polyamide is absorbent with a mildly abrasive surface
Microfiber - 80% polyester, 20% polyamide is highly absorbent and effective wet or dry
The Micronova Slim T™ Mop Covers fit onto the 180° swivel T-Mop™ adapter. Available with an optional thicker foam layer to hold more disinfectant and facilitate wringing. The 305 and 457 mm options are designed for use with the micronova window washing tool for a streak-free finish.
: All components must be ordered separately. For additional Universal Handle Lengths, see Universal Mop Handles from Micronova.
Product Family Options
NovaPoly™ SlimT™ mop covers
Product Information
- LengthMaterialAvailabilityPrice
- MICRT6-10SNovaPoly™ mop cover; sterile
- MICRT6-12SIRNovaPoly™ mop cover irradiated use QPWT-12 adapter; sterileNovaPoly™ mop cover irradiated use QPWT-12 adapter; sterile305 mmPolyester knitLog in for availability1 * 24 Items2.820,00 kr
- MICRT6-12SNovaPoly™ mop cover use QPWT-12 adapter; non irradiatedNovaPoly™ mop cover use QPWT-12 adapter; non irradiated305 mmPolyester knitLog in for availability1 * 1 Items175,00 kr
- MICRT612-14SNovaPoly™ mop cover, thick foam; sterileNovaPoly™ mop cover, thick foam; sterile356 mmPolyester knitLog in for availability1 * 1 Items224,00 kr
- MICRT6-14SNovaPoly™ mop cover, sterile
- MICRT6-18SIRNovaPoly™ mop cover, irradiated use with QPWT-18, QDT-18, QPT-18 adapters; sterileNovaPoly™ mop cover, irradiated use with QPWT-18, QDT-18, QPT-18 adapters; sterile457 mmPolyester knitLog in for availability1 * 24 Items2.970,00 kr
PolySorb™ SlimT™ mop covers
Product Information
- LengthMaterialAvailabilityPrice
- MICRT11-18SIRPolySorb™ mop cover, irradiated use QPWT-18, QDT-18, QPT-18 adapters; irradiatedPolySorb™ mop cover, irradiated use QPWT-18, QDT-18, QPT-18 adapters; irradiated457 mmPolyester knitLog in for availability1 * 24 Items3.090,00 kr
Microfiber SlimT™ mop covers
Product Information
- LengthMaterialAvailabilityPrice
- MICRTAA-12SBlue microfiber mop cover with QPWT-12 adapter; non irradiatedBlue microfiber mop cover with QPWT-12 adapter; non irradiated305 mmPolyester knitLog in for availability1 * 1 Items184,00 kr
- MICRTAW-18SWhite microfiber mop cover use QPWT-18, QDT-18, QPT-18 adapters; non irradiatedWhite microfiber mop cover use QPWT-18, QDT-18, QPT-18 adapters; non irradiated457 mmPolyester knitLog in for availability1 * 1 Items197,00 kr