Order Entry
Denmark
ContactUsLinkComponent
 
Irritant
Corrosive
Hydrogen peroxide 31% VLSI, Electronic Grade
Catalog # 50489142.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Corrosive
Hydrogen peroxide 31% VLSI, Electronic Grade
Catalog # 50489142.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (manganometric):
    30.0 - 32.0 %
  • Colouration:
    ≤ 10 APHA
  • Particle size(>0.50 μm)(/ml)(at filling) (*) :
    ≤ 80
  • Free acid (as H₂SO₄):
    ≤ 40 ppm
  • Non-volatile matter:
    ≤ 20 ppm
  • Cl (Chloride):
    ≤ 500 ppb
  • NH₄ (Ammonium):
    ≤ 1000 ppb
  • NO₃ (Nitrate):
    ≤ 500 ppb
  • PO₄ (Phosphate):
    ≤ 500 ppb
  • SO₄ (Sulphate):
    ≤ 500 ppb
  • Ag (Silver):
    ≤ 10 ppb
  • Al (Aluminium):
    ≤ 10 ppb
  • As (Arsenic):
    ≤ 10 ppb
  • Au (Gold):
    ≤ 20 ppb
  • B (Boron):
    ≤ 20 ppb
  • Ba (Barium):
    ≤ 20 ppb
  • Be (Beryllium):
    ≤ 10 ppb
  • Bi (Bismuth):
    ≤ 10 ppb
  • Ca (Calcium):
    ≤ 50 ppb
  • Cd (Cadmium):
    ≤ 10 ppb
  • Co (Cobalt):
    ≤ 5 ppb
  • Cr (Chromium):
    ≤ 5 ppb
  • Cu (Copper):
    ≤ 5 ppb
  • Fe (Iron):
    ≤ 5 ppb
  • Ga (Gallium):
    ≤ 10 ppb
  • In (Indium):
    ≤ 10 ppb
  • K (Potassium):
    ≤ 50 ppb
  • Li (Lithium):
    ≤ 10 ppb
  • Mg (Magnesium):
    ≤ 5 ppb
  • Mn (Manganese):
    ≤ 10 ppb
  • Mo (Molybdenum):
    ≤ 10 ppb
  • Na (Sodium):
    ≤ 50 ppb
  • Ni (Nickel):
    ≤ 5 ppb
  • Pb (Lead):
    ≤ 10 ppb
  • Pt (Platinum):
    ≤ 10 ppb
  • Sb (Antimony):
    ≤ 10 ppb
  • Sn (Tin):
    ≤ 20 ppb
  • Sr (Strontium):
    ≤ 10 ppb
  • Ti (Titanium):
    ≤ 20 ppb
  • Tl (Thallium):
    ≤ 10 ppb
  • V (Vanadium):
    ≤ 10 ppb
  • Zn (Zinc):
    ≤ 5 ppb
  • Zr (Zirconium):
    ≤ 10 ppb

Specification Test Results

About this item

 :
Boiling Pt:~107 °C (1013 hPa)
Melting Pt:<0 °C
Storage Temperature:Ambient
UN:2014
ADR:5.1 (8),II
 :34.01 g/mol
EINECS:231-765-0
 :GHS07
 :GHS05
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333