Order Entry
Denmark
ContactUsLinkComponent
 
VWR® Nano Spectrophotometer
  76777-152
undefined
VWR® Nano Spectrophotometer
  76777-152

 

  • Model:
    Drop and cuvette
  • Range:
    200 - 800 nm
  • Absorbance range:
    Pedestal: 0,04 - 90 A, Cuvette: 0 - 4 A (10 mm equivalent)
  • Detector:
    2048 Element linear silicone CCD array
  • Light source:
    Pulsed xenon flash lamp
  • Sample size:
    0,5 - 2,0 µl
  • Description:
    VWR® Nano spectrophotometer, EU/UK plug
  • Electrical:
    240 V
  • Wavelength accuracy:
    ±1 nm
  • Output:
    USB and printer
  • Absorbance accuracy:
    1% (7,332 Abs at 260 nm)
  • Concentration:
    Maximum: dsDNA: 4500 ng/µl, BSA: 135 mg/ml
    Lower limit of detection: dsDNA: 2 ng/µl, BSA: 0,1 mg/ml
  • Weight:
    2,8 kg
  • Pk:
    1

 

 

VWR® Nano Spectrophotometer is designed to simplify and accelerate your molecular biology research and routine applications.

  • Rapid calculation and display of 260/280 and 260/230 absorbance ratios
  • Extended-life xenon lamp and monochromator for full spectral scan
  • Auto adjusting pathlengths: 0,2 and 1,0 mm, ensuring accurate measurements
  • Detects up to 4500 ng/µl of undiluted dsDNA
  • Built-in printer and USB port
  • A cuvette port for optical density (OD 600) readings
  • Measurement and data processing time <5 sec
  • Pathlength control adjustment screw driven by electromagnet
  • Standalone instrument control
  • No. of samples: 1

The VWR® Nano Spectrophotometer allows for the swift and accurate purity analysis and quantification of nucleic acids and proteins with as little as 0,5 µL of sample. This stand-alone device operates through a full-colour, 17,8 cm touchscreen with preset programs for nucleic acids and protein sample evaluation, colorimetric assays (BCA, Bradford, Lowry, etc.), UV-Vis spectral scanning, and OD 600 measurements to determine bacteria culture growth. The on-board software includes a useful 'Protocol Guidance' feature to guide the user through each step of the process.

At the heart of the device is a long-life xenon lamp which provides a wide wavelength range from 200 to 800 nm. The xenon lamp system does not require any warm-up time; the instrument can be used immediately upon power-up. A highly sensitive, high-speed silicon CCD detector ensures fast and accurate results.

After pipetting a sample droplet onto the quartz pedestal, the instrument's upper arm is lowered to extend the droplet into a liquid column. The pathlength auto-adjusts from 0,2 to 1 mm, and the absorbance values are evaluated at each to determine the ideal pathlength for optimal concentration calculations. This instrument can rapidly compute DNA, RNA, and protein concentrations as well as purity levels, displaying results within seconds. Users have the option to print the results directly from the instrument’s built-in printer, save to the device’s internal memory, or export the data via USB in an excel-compatible format.

A cuvette port, located behind the quartz pedestal, is used for optical density (OD 600) readings such as bacteria concentration measurements. Two glass cuvettes are included.

 : CE and UKCA marked.
 : Supplied with software, EU plug and UK plug, and 2 glass cuvettes.