Order Entry
Denmark
ContactUsLinkComponent
Peltor™ Z3AF Back Plate
Peltor™ Z3AF Back Plate
Catalog # MMMAPZ3AF
Supplier:  3M
undefined
Peltor™ Z3AF Back Plate
Catalog # MMMAPZ3AF
Supplier:  3M
Specifications
  • Description:
    Back plate kit for Versaflo™ and Speedglas™ series helmets
  • Pk:
    1 KIT
Specifications
About this item
This helmet assembly part securely attaches Peltor™ X-series earmuffs to Versaflo™ headtops M-series or Speedglas™ welding helmet 9100 MP, for effective hearing protection.
  • Attach earmuffs to a headtop or helmet
  • Check equipment regularly to increase service life
Ensure effective hearing protection when wearing a headtop or helmet with helmet assembly part Z3AF/2. This assembly part is designed to attach 3M™ Peltor™ X-series earmuffs to 3M™ Versaflo™ headtop M-series or 3M™ Speedglas™ welding helmet 9100 MP.