Cole-Parmer® HM-200 Heating Mantles
Supplier: Cole-Parmer
Certificates
About this item
Thermal insulating cartridge provides maximum heat transfer.
- Chemical resistant polypropylene housing is 'safe to the touch'
Quickly and safely heat round-bottom flasks to 450 °C (842 °F). These simple-to-use heating mantles feature a built-in power controller with dial markings from 1 to 10 for repeatability. An amber coloured 'heater on' light and a clear-coloured 'power on' light indicate when the heating mantle is in use. The replaceable coiled heating element suspended within a thermal insulating cartridge provides maximum heat transfer and support.
Ventilation slots and ceramic fibre insulation keep the chemical resistant polypropylene exterior cool even while in use. A safety earth (ground) screen and double fuses provide added safety.
Specifications
- Catalog No:
- ELET36002-26
- ELET36002-22
- ELET36002-18
- ELET36002-06
- ELET36002-14
- ELET36002-10
- ELET36002-30
- ELET36002-02
- Electrical:
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- 230 V, 50/60 Hz
- Max. temperature:
- 450 °C
- 450 °C
- 450 °C
- 450 °C
- 450 °C
- 450 °C
- 450 °C
- 450 °C