DURAN® Safety Joints, Cone with Rim
Supplier: DWK Life Sciences
Certificates
About this item
Cones with collar and additionally moulded glass ring.
- Made from borosilicate 3.3 glass
- Cone with collar and additionally moulded glass ring prevents unintentional loosening of the ground joint
- Firmly seated ground joints can be loosened more easily here, the danger of glass breakage and associated risks is thus minimised
- Use in equipment for handling hazardous substances or working with pressure differences in the high vacuum range
- Tight closure due to standard ground joint
In order to meet high safety demands, example when dealing with hazardous substances or pressure differences, DURAN® Safety Joints offer very good protection against accidental loosening of the connection. Furthermore, DURAN® Safety Joints offer a system to separate jammed cones and sockets. This minimises the risk of breakage and related threats.
Frequently Bought Together