Order Entry
Denmark
ContactUsLinkComponent
Orion™ AquaMate Vis and UV-Vis Spectrophotometers
Orion™ AquaMate Vis and UV-Vis Spectrophotometers
Catalog # ORIOAQ8100
Supplier:  Thermo Orion
undefined
Orion™ AquaMate Vis and UV-Vis Spectrophotometers
Catalog # ORIOAQ8100
Supplier:  Thermo Orion

Specifications

  • Detector:
    Detector type: Dual silicon photodiodes
  • Length:
    35,5 cm
  • Description:
    AquaMate 8100 UV-Vis spectrophotometer with north america, european and UK power cord
  • Electrical:
    External AC to DC converter. Voltage and frequency (Hz) selected automatically, 100/240 V, 50/60 Hz
  • Width:
    38,5 cm
  • Depth:
    35,5 cm
  • Wavelength range:
    190 - 1100 nm
  • Wavelength accuracy:
    ±0,5 nm
  • Cell compartment:
    Accessible from top, front or side Able to accommodate cells up to 100 mm cells Available accessory for test tubes up to 25 mm diameter and 150 mm height removable, washable sample compartment liner with magnetic placement and hold-down
  • Display:
    7" High definition color touchscreen, fixed, 800×1280 pixels
  • Drift:
    <0,0005 A/h (at 500 nm after 1 h warmup)
  • Height:
    19,5 cm
  • Interfaces:
    Glove friendly touch-screen
  • Lamp source, lifetime:
    >5 years typical
  • Noise:
    (RMS at 500 nm 60 consecutive measurements) ≤0,00020 A at 0 A at 260 and 500 nm ≤0,00030 A at 1 A at 260 and 500 nm ≤0,00040 A at 2A at 260 and 500 nm
  • Optical design:
    Double beam
  • Photometric accuracy:
    ±0,002 A at 0,5 A, ±0,004 A at 1 A, ±0,008 A at 2 A
  • Photometric range:
    −2 - +3,5 A
  • Printer:
    Snap-on printer included
  • Scan speed:
    Slow, medium and fast up to 1600 nm/min
  • Spectral bandwidth:
    2 nm
  • Stray light:
    <1,0%T 198 nm (KCl) , <0,05%T at 220 nm (NaI), <0,03%T at 340 nm (NaNO₂)
  • Wavelength repeatability:
    <±0,2 nm
  • Weight:
    7,5 kg
  • Pk:
    1
  • Type:
    UV-Vis spectrophotometer
  • For:
    Water and waste water analysis

Specifications

About this item

Thermo Scientific™ Orion™ AquaMate UV-Vis spectrophotometers include over 260 preprogramed methods, and are designed to meet the needs of water and wastewater analysis laboratories. AquaMate UV-Vis and Visible-only spectrophotometers incorporate advanced features and functions for ultimate flexibility in operation.

  • More than 260 preprogramed methods offer easy and convenient measurements using Orion AQUAfast™, Merck™ or CHEMetrics™ chemistries
  • Preprogramed methods can also be adapted for new chemistries or operators can create their own custom methods and calibration curves
  • Instruments allow a one-point adjustment on any preprogramed method to correct for variations in batch reagent chemistries
  • Glove friendly 7" touch screen controller
  • Performance verification tests ensure wavelength accuracy and instrument functionality
  • 1 year warranty

Wide variety of vials sizes can be used, including 10 mm square vials, 20 and 50 mm rectangular vials and an adjustable round vial holder that accommodates vials and test tube from 12 to 25 mm OD.

Each verification report gives the time, date and instrument serial number in accordance with GLP and GMP. Built-in wavelength accuracy test is compatible with either the internal lamp or external calibrated standards. Built-in test methods for stray light, noise and resolution are available to validate instrument performance further.

Baseline flatness: ±0,002 A
Connections: Single USB-AExport data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, availablePrint via USB, Ethernet or Wi-Fi USB adaptor available
Photometric display: −3 to +5 A
Photometric repeatability: ±0,001 A at 1 A measured at 1 A at 546 nm
Wavelength data interval: 0,2, 0,5, 1, 2, 5 nm

Certifications: CE/TUV.
Delivery: Test tube holder for vials and test tube from 12 to 25 mm OD, 10 mm square single-cell holder, AquaMate water analysis methods software, snap-on printer, dust cover, external power supply, north america power cable (NEMA 5-15), european power cable (CEE 7/7), UK power cable (BS 1362).