Order Entry
Denmark
ContactUsLinkComponent
b.safe Blind Fitting PFAS Analytic
b.safe Blind Fitting PFAS Analytic
Catalog # BOHLM401-01
Supplier:  Bohlender
undefined
b.safe Blind Fitting PFAS Analytic
Catalog # BOHLM401-01
Supplier:  Bohlender
Specifications
  • Thread:
    UNF ¹/₄"
  • Description:
    Blind fitting PFAS analytic, 10 per pack
  • Pk:
    1
Specifications
About this item
To close unused connections on b.safe Caps PFAS Analytic.
  • Made of PE
  • For PFAS analysis
  • With ergonomically shaped grip surface
  • easy mounting
Blind Fittings made of PE for tight closure of unused connectors on b.safe Caps PFAS Analytic. With ergonomic gripping surface for easy assembly.