Order Entry
Denmark
ContactUsLinkComponent
Printer Ribbons, M710 R4400 Series
Printer Ribbons, M710 R4400 Series
Catalog # BRDY173192
Supplier:  Brady
undefined
Printer Ribbons, M710 R4400 Series
Catalog # BRDY173192
Supplier:  Brady
Specifications
  • Colour:
    White
  • Description:
    M710 R4400 series printer ribbon for M710 and BMP71 printers
  • Length:
    45.72 m
  • Environmentally Preferable:
  • Pk:
    1
  • Width:
    50 mm
  • For:
    M710;BMP71
Specifications
About this item
One-color ribbon for printing a single color onto a label.
  • R4400 series resin formulation offers excellent resistance to solvents, scratches, and high heat when printed on the appropriate Brady® label material
  • Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra aggressive polyester and B-423 permanent polyester
  • Self-contained M710 series ribbon cartridges insert straight into the printer with ease
M710 Series snap-in print ribbons enable 5 second ribbon supply changes. No calibrating, no threading, no waste, no hassles, and the ink always faces the right direction.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability.