Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cocamidopropyl betaine
Cocamidopropyl betaine solution
Cocamidopropyl betaine solution
Irritant
Cocamidopropyl betaine solution
Catalog # SPCMC3493-200LTBL
Supplier:
Spectrum Chemical
CAS Number:
Expand image
Irritant
Cocamidopropyl betaine solution
Catalog # SPCMC3493-200LTBL
Supplier:
Spectrum Chemical
CAS Number:
Specifications
Pk:
200 L
Seq:
5549
Specifications
About this item
Product Details
:
:
GHS07
Storage Temperature:
Ambient
CAS Number:
61789-40-0
Product Details