
Iron(II) sulphate hydrate, dried, powder USP
Catalog # SPCMF1065-2.5KG
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Poly pail
- Pk:2,5 kg
- Seq:4076
Specifications
Specification Test Results
- Assay:86% Min.
Specification Test Results
About this item
:
:169.93 g/mol
Melting Pt:∼300 °C
Density:2,97 g/cm³ (20 °C)
EINECS:231-753-5
:GHS07
Storage Temperature:Ambient
CAS Number:13463-43-9
Formula:FeSO₄·H₂O
MDL Number:MFCD00149722