Squalene ≥99%
Poly pail, 5 L
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Squalene, is an organic compound that be found occurring naturally in shark liver oil and certain plants. It is a hydrocarbon that all plants and animals produce in varying quantities. Mostly used in cosmetics as a natural skin moisturiser, it has recently been used as an immunologic adjuvant in certain vaccines.
Synonyms: 2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene
- CAS Number:111-02-4
- Formula:C₃₀H₅₀
- MDL Number:MFCD00008912
- Molecular Weight:410.73 g/mol
- Merck Index:13,08847
- Boiling Pt:429 °C (1013 hPa)
- Melting Pt:–5 °C
- Density:0,858
- Flash Pt:200 °C
- Storage Temperature:Ambient
- EINECS:203-826-1
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Assay:99% Min.
Specifications
- Pack type:Poly pail
- Pk:5 L



