Order Entry
Denmark
ContactUsLinkComponent
Adipohydrazide ≥99%
Adipohydrazide ≥99%
Catalog # SPCMA3802-20KGBL
Supplier:  Spectrum Chemical
CAS Number:  
Adipohydrazide ≥99%
Catalog # SPCMA3802-20KGBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    20 kg
  • Seq:
    4241
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    99% Min.

Specification Test Results

About this item

Synonyms: Adipyl hydrazide
 :
 :174.2 g/mol
Boiling Pt:446 °C (1013 hPa)
Melting Pt:181…182 °C
Storage Temperature:Ambient
EINECS:213-999-5
CAS Number:1071-93-8
Formula:C₆H₁₄N₄O₂
MDL Number:MFCD00007614