Order Entry
Denmark
ContactUsLinkComponent
 
Magnesium sulphate, anhydrous ≥99.0% USP
Catalog # SPCMM1354-125GM
Supplier:  Spectrum Chemical
CAS Number:  
undefined
Magnesium sulphate, anhydrous ≥99.0% USP
Catalog # SPCMM1354-125GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    125 g
  • Seq:
    3445

Specifications

Specification Test Results

  • Assay (Anhydrous Basis):
    99.0 - 100.5%
  • pH (In Solution 1 in 20):
    5.0 - 9.2
  • Loss On Drying:
    max. 2 %
  • Chloride (Cl):
    max. 0.014 %
  • Iron:
    max. 20 Ug/G

Specification Test Results

About this item

 :
 :120.37 g/mol
Storage Temperature:Ambient
CAS Number:7487-88-9
Formula:MgSO₄
MDL Number:MFCD00011110