Hydroxyurea ≥97% USP
About this item
Hydroxyurea, USP is an antineoplastic drug used in myeloproliferative disorders. It has also been considered useful in treating painful attacks in sickle-cell disease and diseases like AIDS. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
- CAS Number:127-07-1
- Formula:CH₄N₂O₂
- MDL Number:MFCD00007943
- Molecular Weight:76.05 g/mol
- Merck Index:13,04874
- Boiling Pt:296 °C (1013 hPa)
- Melting Pt:142…144 °C
- Storage Temperature:Ambient
- EINECS:204-821-7
5 Options Available Below
Product Details & Documents
Hydroxyurea, USP is an antineoplastic drug used in myeloproliferative disorders. It has also been considered useful in treating painful attacks in sickle-cell disease and diseases like AIDS. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
- CAS Number:127-07-1
- Formula:CH₄N₂O₂
- MDL Number:MFCD00007943
- Molecular Weight:76.05 g/mol
- Merck Index:13,04874
- Boiling Pt:296 °C (1013 hPa)
- Melting Pt:142…144 °C
- Storage Temperature:Ambient
- EINECS:204-821-7
Specifications for Product Family
Specification Test Results
- Assay:97% Min.
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



