Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium hydroxide ≥95.0%, pellets NF

Sodium hydroxide ≥95.0%, pellets NF

Poly bottle, 500 g
Catalog # SPCMSO170-500GM
Corrosive
Irritant
Supplier:   Spectrum Chemical
Sodium hydroxide ≥95.0%, pellets NF
Catalog # SPCMSO170-500GM
Supplier:  Spectrum Chemical
CAS Number:  1310-73-2
Sodium hydroxide ≥95.0%, pellets NF

Poly bottle, 500 g

CAS Number:  1310-73-2
Price
982,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Sodium Hydroxide, Pellets, NF is used as a pH adjusting agent.

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Storage Temperature:Ambient
  • UN:1823
  • ADR:8,II
Specifications

Specification Test Results

  • Assay (Total Alkali Including A:
    95.0 - 100.5%
  • Insoluble Substances & Organic Matter:
    To Pass Test
  • Potassium (K):
    To Pass Test
  • Heavy Metals:
    max. 30 ppm

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.