Order Entry
Denmark
ContactUsLinkComponent
 
Anti-DARS2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-14198R-A750
 :  Bioss
Anti-DARS2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-14198R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    DARS2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Epitope:
    551-645/645
  • Form:
    Liquid
  • Gene ID:
    55157
  • Antigen synonyms:
    ASPRS|Aspartyl-tRNA synthetase|LBSL|Aspartate--tRNA ligase, mitochondrial|MT-ASPRS|DARS2
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    68 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human DARS2
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

ATP + L-aspartate + tRNA(Asp) = AMP + diphosphate + L-aspartyl-tRNA(Asp).

Type: Primary
Antigen: DARS2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 551-645/645
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat