Specifications
- Antibody type:Primary
- Antigen name:MPP7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:301-400/576
- Form:Liquid
- Antigen synonyms:FLJ32798|MAGUK p55 subfamily member 7|Palmitoylated membrane protein 7|Membrane protein palmitoylated 7 MAGUK p55 subfamily member 7
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:65 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human MPP7
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The MAGUK (membrane-associated guanylate kinase homologs) family of proteins contain multiple protein-binding domains and are involved in cell junction organization, tumor suppression, and signaling. The MAGUK family is divided into four subfamilies: DLG-like, ZO1-like, p55-like and LIN2-like. MPP7 (MAGUK p55 subfamily member 7), also known as palmitoylated membrane protein 7, is a 576 amino acid protein that belongs to the MAGUK family and assists in the assembly of protein complexes. A peripheral membrane protein that exists as a heterodimer, MPP7 enhances tight junction formation and epithelial cell polarity. MPP7 contains one SH3 domain, a guanylate kinase-like domain, two L27 domains and a single PDZ (DHR) domain. The gene encoding MPP7 maps to human chromosome 10p11.23
Type: Primary
Antigen: MPP7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 301-400/576
Host: Rabbit
Isotype: IgG
Reactivity: