Anti-MS4A6A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-13692R-A680
: Bioss
- Antibody type:Primary
- Antigen name:MS4A6A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:51-150/248
- Form:Liquid
- Antigen synonyms:4SPAN3|CDA01|CD20L3|MST090|Four span transmembrane protein 32|CD20 like precusor|CD20 antigen like 3|Membrane spanning 4 domains subfamily A member 6A|MSTP090|MS4A6A polymorph|HAIRB iso|Four span transmembrane protein 3|Four span transmembrane protein 31|4SPAN3.2|MS4A6
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:27 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human MS4A6A
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
MS4A6A is a member of the membrane-spanning 4A gene family and may be involved in signal transduction as a component of a multimeric receptor complex. There are three named isoforms.
Type: Primary
Antigen: MS4A6A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/248
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat