Anti-DNA Ligase IV/LIG4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-1677R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:DNA Ligase IV/LIG4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:51-150/911
- Form:Liquid
- Gene ID:3981
- Antigen synonyms:DNA ligase IV|Polydeoxyribonucleotide synthase [ATP] 4|LIG4S|DNA ligase 4|LIG4
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:104 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human DNA Ligase IV
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Efficiently joins single-strand breaks in a double-stranded polydeoxynucleotide in an ATP-dependent reaction. Involved in DNA non-homologous end joining (NHEJ) required for double-strand break repair and V(D)J recombination. The LIG4-XRCC4 complex is responsible for the NHEJ ligation step, and XRCC4 enhances the joining activity of LIG4. Binding of the LIG4-XRCC4 complex to DNA ends is dependent on the assembly of the DNA-dependent protein kinase complex DNA-PK to these DNA ends.
Type: Primary
Antigen: DNA Ligase IV/LIG4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/911
Host: Rabbit
Isotype: IgG
Reactivity: Human