Order Entry
Denmark
ContactUsLinkComponent
Anti-Complement C3 Goat Polyclonal Antibody (Alexa Fluor® 680)
Anti-Complement C3 Goat Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-0367G-A680
Supplier:  Bioss
Anti-Complement C3 Goat Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-0367G-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Complement C3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    718
  • Antigen synonyms:
    Complement C3 beta chain|AHUS5|Plp.|Complement C3c alpha' chain fragment 2|ARMD9|Complement component 3 precursor|Complement component 3|Complement C3c fragment|Complement C3b alpha chain|Complement C3d fragment|Complement C3|Complement C3g fragment|Complement C3 precursor|C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1 antibody C3 antibody C3a anaphylatoxin|Complement component C3|ASP|CPAMD1|Complement C3f fragment|Acylation-stimulating protein cleavage product|CO3_HUMAN|Complement C3 alpha chain|Complement factor 3|Acylation stimulating protein cleavage product
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    187 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    human Complement C3 purified from human plasma
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

The complement factor C3 consists of an alpha and a beta chain. C3 is a central factor in the complement cascade. It is central to the alternative pathway that leads to the C3 convertase C3bBb. The classical mannose binding lectin activation pathway leads to the C3 convertase C4b2a. These convertases cleave C3 resulting in C3a and C3b. Further degradation leads to the formation of the alpha chain products C3d, C3g and C3c. C3 is an acute phase protein that is produced by a wide range of tissues, including renal epithelial cells and hepatocytes.

Type: Primary
Antigen: Complement C3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Goat
Isotype: IgG
Reactivity: Mouse