Specifications
- Antibody type:Primary
- Antigen name:HBeAg
- Clonality:Monoclonal
- Clone:2E9
- Conjugation:Alexa Fluor® 750
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Virus
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:External core antigen|HBeAg|Core protein|Hepatitis B|HBe antigen|Precore/core|Precore protein|Core antigen|Pre C,C|Precore/core protei|HBVgp4|HBEAG_HBVD3|Precore/core ORF|p25
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:HBeAg full length protein
- Purification:Purified by Protein G
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Hepatitis B e-antigen (HBeAg) is a viral protein associated with HBV infections. Unlike the surface antigen, the e-antigen is found in the blood only when there are viruses also present. When the virus goes into hiding,? the e-antigen will no longer be present in the blood. HBeAg is often used as a marker of ability to spread the virus to other people (infectivity). Measurement of e-antigen may also be used to monitor the effectiveness of HBV treatment; successful treatment will usually eliminate HBeAg from the blood and lead to development of against e-antigen (anti-HBe). There are some types (strains) of HBV that do not make e-antigen; these are especially common in the Middle East and Asia. In areas where these strains of HBV are common, testing for HBeAg is not very useful.
Type: Primary
Antigen: HBeAg
Clonality: Monoclonal
Clone: 2E9
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Mouse
Isotype: IgG
Reactivity: Virus