Anti-GAL3ST4 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
BOSSBS-16215R-A680
: Bioss
- Antibody type:Primary
- Antigen name:GAL3ST4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:401-486/486
- Form:Liquid
- Gene ID:79690
- Antigen synonyms:Galactose-3-O-sulfotransferase 4|Beta-galactose-3-O-sulfotransferase 4|GAL3ST-4|Gal-beta-1,3-GalNAc 3'-sulfotransferase|GAL3ST4|PP6968
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:54 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GAL3ST4
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Catalyses the transfer of sulfate to beta-1,3-linked galactose residues in O-linked glycoproteins. Good substrates include asialofetuin, Gal-beta-1,3-GalNAc and Gal-beta-1,3 (GlcNAc-beta-1,6)GalNAc.
Type: Primary
Antigen: GAL3ST4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-486/486
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat